Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000554706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007501.hg.1 |
ENST00000267584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007501.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007501.hg.1 |
TC14000604.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400157690.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157690.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157690.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157690.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157690.hg.1 |
TC1400008118.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000267584.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000554706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000555570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
ENST00000556643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008118.hg.1 |
NM_152327.4 |
Refseq |