Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004400.hg.1 |
TC01000286.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004400.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004400.hg.1 |
ENST00000374639.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004400.hg.1 |
ENST00000374640.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004400.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004400.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150778.hg.1 |
TC0100007291.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374639.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
ENST00000374640.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_001114101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007291.hg.1 |
NM_172369.4 |
Refseq |