Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198274.3 | NP_938015.1 | 5 - 6 | 93 | ||
GenBank mRNA | AK091724.1 | - | 5 - 6 | 698 | 93 | |
AL832035.1 | - | 5 - 6 | 93 | |||
AY518933.1 | - | 5 - 6 | 93 | |||
BC126191.1 | - | 5 - 6 | 8991907 | 93 | ||
BX647878.1 | - | 5 - 6 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |