Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042742.hg.1 |
TC01002725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042742.hg.1 |
ENST00000371153.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042742.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042743.hg.1 |
TC01002725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042743.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042743.hg.1 |
ENST00000371153.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042743.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042745.hg.1 |
TC01002725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042745.hg.1 |
ENST00000371153.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042745.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042744.hg.1 |
TC01002725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042744.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042744.hg.1 |
ENST00000371153.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042744.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207834.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207834.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207834.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207834.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207834.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207834.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207834.hg.1 |
TC0100014403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000317477.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371150.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014403.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207832.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207832.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207832.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207832.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207832.hg.1 |
TC0100014403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100207836.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207836.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207836.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207836.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207836.hg.1 |
TC0100014403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100207835.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207835.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207835.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207835.hg.1 |
ENST00000354381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207835.hg.1 |
ENST00000371153.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207835.hg.1 |
NM_181712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207835.hg.1 |
TC0100014403.hg.1 |
Clariom D TC |