Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007822.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007822.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007822.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007822.hg.1 |
TC14000636.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158425.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158425.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158425.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158425.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158425.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158425.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158425.hg.1 |
TC1400008209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000361529.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
ENST00000557052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008209.hg.1 |
NM_207117.3 |
Refseq |