Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013202.hg.1 |
TC19002670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013202.hg.1 |
ENST00000196548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013202.hg.1 |
ENST00000438500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013202.hg.1 |
ENST00000546178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013202.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013208.hg.1 |
TC19002670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013208.hg.1 |
ENST00000196548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013208.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203245.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203245.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203245.hg.1 |
TC1900011827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203245.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203245.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203248.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203248.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203248.hg.1 |
TC1900011827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203248.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203248.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011827.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203246.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203246.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203246.hg.1 |
TC1900011827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203246.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203246.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203247.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203247.hg.1 |
NM_021089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203247.hg.1 |
TC1900011827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203247.hg.1 |
ENST00000621650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203247.hg.1 |
NM_021089.2 |
Refseq |