Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010508.hg.1 |
TC06000890.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010508.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010508.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010507.hg.1 |
TC06000890.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010507.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010507.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010505.hg.1 |
TC06000890.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010505.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162794.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162794.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162794.hg.1 |
TC0600009144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009144.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162797.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162797.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162797.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162797.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162797.hg.1 |
TC0600009144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600162799.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162799.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162799.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162799.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162799.hg.1 |
TC0600009144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600162796.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162796.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162796.hg.1 |
ENST00000368847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162796.hg.1 |
NM_153369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162796.hg.1 |
TC0600009144.hg.1 |
Clariom D TC |