Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000349394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000555154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000555154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
ENST00000556415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007864.hg.1 |
NM_007224.3 |
Refseq |