Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017564.hg.1 |
TC02001161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017564.hg.1 |
ENST00000357799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017564.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017564.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017564.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017564.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000460534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000460534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000460534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000418045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000460534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000460534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000460534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000469840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
ENST00000488636.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010419.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
ENST00000357799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
ENST00000409203.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
NM_001160033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
NM_001160046.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
NM_152524.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200173526.hg.1 |
TC0200010419.hg.1 |
Clariom D TC |