Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006545.hg.1 |
ENST00000315066.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006545.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006545.hg.1 |
TC17000537.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157759.hg.1 |
TC1700007916.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000315066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000585355.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000586652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000588527.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000591559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
ENST00000592166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007916.hg.1 |
NM_173478.2 |
Refseq |