Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022827.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022827.hg.1 |
ENST00000368854.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022827.hg.1 |
ENST00000418782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022827.hg.1 |
ENST00000474277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022827.hg.1 |
ENST00000540123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022827.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10022827.hg.1 |
TC10001721.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000481775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012108.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
ENST00000241305.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
ENST00000368854.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
ENST00000615851.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
NM_198148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000182817.hg.1 |
TC1000012108.hg.1 |
Clariom D TC |