Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015149.hg.1 |
TC0X001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015149.hg.1 |
ENST00000239666.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015149.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015149.hg.1 |
ENST00000473667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015149.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009602.hg.1 |
ENST00000554427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015148.hg.1 |
TC0X001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015148.hg.1 |
ENST00000239666.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015148.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015148.hg.1 |
ENST00000473667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015148.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015150.hg.1 |
TC0X001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015150.hg.1 |
ENST00000239666.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015150.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015150.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015145.hg.1 |
TC0X001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015145.hg.1 |
ENST00000239666.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015145.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015145.hg.1 |
ENST00000473667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015145.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009964.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009602.hg.1 |
ENST00000554427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168025.hg.1 |
TC0X00009964.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
ENST00000486461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168021.hg.1 |
TC0X00009964.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
ENST00000473667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168026.hg.1 |
TC0X00009964.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00168027.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168027.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168027.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168027.hg.1 |
ENST00000239666.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168027.hg.1 |
ENST00000374454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168027.hg.1 |
NM_016484.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168027.hg.1 |
TC0X00009964.hg.1 |
Clariom D TC |