Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019202.hg.1 |
TC04001373.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019202.hg.1 |
ENST00000295908.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019202.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000363599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000509340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000509340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000509340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000511506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000511506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000511506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177170.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177170.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177170.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177170.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177170.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177170.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177170.hg.1 |
TC0400011283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000295908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000363599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000505022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000506423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000508256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000509340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000509340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000509340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000510548.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000511506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000511506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000511506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000513546.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000514204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
ENST00000608933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011283.hg.1 |
NM_152542.4 |
Refseq |