Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014629.hg.1 |
TC0X001073.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014629.hg.1 |
ENST00000354646.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014629.hg.1 |
ENST00000375159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014629.hg.1 |
ENST00000375169.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014629.hg.1 |
NM_001002838.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014629.hg.1 |
NM_020922.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000354646.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000375159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000375169.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000620763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
NM_001002838.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
NM_020922.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000354646.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000375159.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000375169.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
ENST00000620763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
NM_001002838.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
NM_020922.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166987.hg.1 |
TC0X00009778.hg.1 |
Clariom D TC |