Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000565930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000565930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
ENST00000565930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011700.hg.1 |
ENST00000267892.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011700.hg.1 |
ENST00000356231.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011700.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011700.hg.1 |
TC15001268.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500166881.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500166881.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500166881.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500166881.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500166881.hg.1 |
TC1500009199.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000356231.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000562465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000563604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000565930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000565930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
ENST00000565930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009199.hg.1 |
NM_138477.3 |
Refseq |