Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020892.hg.1 |
TC08001599.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020892.hg.1 |
ENST00000297632.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020892.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020896.hg.1 |
TC08001599.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020896.hg.1 |
ENST00000297632.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020896.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020893.hg.1 |
TC08001599.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020893.hg.1 |
ENST00000297632.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020893.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020895.hg.1 |
TC08001599.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020895.hg.1 |
ENST00000297632.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020895.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020894.hg.1 |
TC08001599.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020894.hg.1 |
ENST00000297632.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020894.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188055.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188055.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188055.hg.1 |
TC0800012457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188052.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188052.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188052.hg.1 |
TC0800012457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188053.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188053.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188053.hg.1 |
TC0800012457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188056.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188056.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188056.hg.1 |
TC0800012457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188054.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188054.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188054.hg.1 |
TC0800012457.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188057.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188057.hg.1 |
NM_194291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188057.hg.1 |
TC0800012457.hg.1 |
Clariom D TC |