Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005268467.1 | XP_005268524.1 | 8 - 9 | 1500 | 130 | |
XM_005268468.1 | XP_005268525.1 | 8 - 9 | 1497 | 130 | ||
GenBank mRNA | AK128738.1 | - | 8 - 9 | 1497 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |