Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002782.hg.1 |
ENST00000325192.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002782.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002782.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002782.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002782.hg.1 |
TC14000261.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007029.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000325192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000553841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000555826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
ENST00000557112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
NM_001017923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148784.hg.1 |
TC1400007029.hg.1 |
Clariom D TC |