Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009991.hg.1 |
TC06000846.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009991.hg.1 |
ENST00000285105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009991.hg.1 |
ENST00000369066.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009991.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161916.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161916.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161916.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161916.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161916.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161916.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161916.hg.1 |
TC0600008981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000369066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000457437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000487681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
ENST00000633556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008981.hg.1 |
NM_001624.3 |
Refseq |