Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030805.hg.1 |
TC07002016.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030805.hg.1 |
ENST00000223271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030805.hg.1 |
ENST00000466675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030805.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030805.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030804.hg.1 |
TC07002016.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030804.hg.1 |
ENST00000223271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030804.hg.1 |
ENST00000466675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030804.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030804.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007882.hg.1 |
ENST00000474487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010146.hg.1 |
ENST00000567349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013027.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007882.hg.1 |
ENST00000474487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193856.hg.1 |
TC0700013027.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000223271.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000466675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000467793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000478771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
ENST00000482669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
NM_002889.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193855.hg.1 |
TC0700013027.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010146.hg.1 |
ENST00000567349.1 |
Ensembl |