Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004795.hg.1 |
TC0X000372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004795.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004795.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151576.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151576.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151576.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151576.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151576.hg.1 |
TC0X00007532.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000374521.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
ENST00000480702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007532.hg.1 |
NM_001013579.2 |
Refseq |