Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011378.hg.1 |
TC06000970.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011378.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011378.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164514.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164514.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164514.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164514.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164514.hg.1 |
TC0600009406.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000329722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
ENST00000498112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009406.hg.1 |
NM_001010905.2 |
Refseq |