Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010431.hg.1 |
ENST00000540082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000366607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000467767.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
ENST00000473132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017815.hg.1 |
NM_014765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010431.hg.1 |
ENST00000540082.1 |
Ensembl |