Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005620.hg.1 |
ENST00000399154.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005620.hg.1 |
ENST00000457912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005620.hg.1 |
ENST00000527972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005620.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005620.hg.1 |
TC11000466.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100153996.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153996.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153996.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153996.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153996.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153996.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153996.hg.1 |
TC1100007682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000399154.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
ENST00000527972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007682.hg.1 |
NM_001105565.2 |
Refseq |