Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000456958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000456958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000456958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000601406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000601406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000601829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000601829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013912.hg.1 |
ENST00000538831.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002045.hg.1 |
ENST00000456958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002045.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002045.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002045.hg.1 |
TC19000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002044.hg.1 |
ENST00000456958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002044.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002044.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002044.hg.1 |
TC19000131.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000601406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000601829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
TC1900011655.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000601406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000601829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199188.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000456958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000456958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000456958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000594797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000598540.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000600836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000601406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000601406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000601829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000601829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
ENST00000623154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NM_001171155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011655.hg.1 |
NR_033242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013912.hg.1 |
ENST00000538831.2 |
Ensembl |