Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016680.hg.1 |
ENST00000334258.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016680.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016680.hg.1 |
ENST00000554873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016680.hg.1 |
ENST00000557275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016680.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14016680.hg.1 |
TC14001476.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400183272.hg.1 |
TC1400010784.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000334258.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000553340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000554873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
ENST00000555759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010784.hg.1 |
NM_152592.4 |
Refseq |