Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267869.1 | NP_001254798.1 | 1 - 2 | 506 | 128 | |
NM_198704.2 | NP_941993.1 | 1 - 2 | 506 | 128 | ||
NM_198707.2 | NP_941996.1 | 1 - 2 | 506 | 128 | ||
GenBank mRNA | AY268353.1 | - | 1 - 2 | 230 | 128 | |
AY313895.1 | - | 1 - 2 | 247 | 128 | ||
BC094798.1 | - | 1 - 2 | 262 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |