Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000292778.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000398873.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000415762.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000464015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000468686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000473985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
ENST00000482998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008099.hg.1 |
NM_001017964.1 |
Refseq |