Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009532.hg.1 |
TC03000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009532.hg.1 |
ENST00000394140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009532.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000608041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000608041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
ENST00000608041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009533.hg.1 |
TC03000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009533.hg.1 |
ENST00000394140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009533.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009531.hg.1 |
TC03000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009531.hg.1 |
ENST00000394140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009531.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009536.hg.1 |
TC03000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009536.hg.1 |
ENST00000394140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009536.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009535.hg.1 |
TC03000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009535.hg.1 |
ENST00000394140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009535.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009538.hg.1 |
TC03000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009538.hg.1 |
ENST00000394140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009538.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009534.hg.1 |
TC03000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009534.hg.1 |
ENST00000394140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009534.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000608041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000608041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
ENST00000608041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008144.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000394140.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000460317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000465368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000472392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000478856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000480073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
ENST00000497785.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
NM_020202.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158023.hg.1 |
TC0300008144.hg.1 |
Clariom D TC |