Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008778.1 | NP_001008778.1 | 5 - 6 | 519 | 113 | |
XM_017017706.1 | XP_016873195.1 | 4 - 5 | 913 | 113 | ||
XM_017017708.1 | XP_016873197.1 | 4 - 5 | 913 | 113 | ||
GenBank mRNA | AY820305.1 | - | 5 - 6 | 519 | 113 | |
BC137244.1 | - | 5 - 6 | 601 | 9021241 | 113 | |
BC137246.1 | - | 5 - 6 | 601 | 9021243 | 113 | |
BC144548.1 | - | 5 - 6 | 601 | 9053075 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255041 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842253 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842254 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842257 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255042 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842261 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842236 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842243 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255019 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842262 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842239 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842241 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255052 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842244 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255045 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842255 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842249 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842260 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255040 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28255035 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842258 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07842237 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 953487 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3322945 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0794 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0793 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3240 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02458614 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P56648234 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02458611 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3322945 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3322945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842241 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255045 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255028 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842230 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255041 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842231 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842245 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842244 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842240 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255042 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842237 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255024 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842245 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255042 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255028 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255045 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842231 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255041 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842240 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842244 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842230 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842241 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28255024 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07842237 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02458611 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P493262 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842228 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842240 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842237 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842259 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842274 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842275 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16462103 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842245 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16462095 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842243 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842272 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842269 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842251 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842235 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842230 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842271 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842278 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842257 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16462099 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842233 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842255 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842241 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16462108 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842277 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16462101 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07842262 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001008778_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7941174 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7941175 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7941178 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7941177 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7941176 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7941173 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7941172 | - | - |
Affymetrix Human Genome U219 Array | 11735777_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11735777_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11007662.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007662.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11007662.hg.1 | TC11000619.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11007661.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007661.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11007661.hg.1 | TC11000619.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100157617.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100157617.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100157617.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100157617.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100157617.hg.1 | TC1100008004.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008004.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100157616.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100157616.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100157616.hg.1 | ENST00000377185.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100157616.hg.1 | NM_001008778.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100157616.hg.1 | TC1100008004.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046579 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046579 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 174068 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777476 | AY820305.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777476 | BC137244.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777476 | BC137246.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_1637-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 3890255 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_1637 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1777476 | BC144548.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1777476 | NM_001008778.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1777476 | AY820305.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1777476 | BC137244.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1777476 | BC137246.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1777476 | BC144548.1 | human_transcriptome |