Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004197.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004197.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004197.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004197.hg.1 |
TC14000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004196.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004196.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004196.hg.1 |
TC14000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004201.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004201.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004201.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004201.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004201.hg.1 |
TC14000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151572.hg.1 |
TC1400007396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151567.hg.1 |
TC1400007396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000554895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000555934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007396.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000334389.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000553426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000623219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
ENST00000623985.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
NR_015358.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151568.hg.1 |
TC1400007396.hg.1 |
Clariom D TC |