Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008470.hg.1 |
ENST00000419457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008470.hg.1 |
ENST00000419457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024107.hg.1 |
TC07003361.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024107.hg.1 |
ENST00000003100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024107.hg.1 |
ENST00000450723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024107.hg.1 |
ENST00000496998.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024107.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024107.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011670.hg.1 |
ENST00000492267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024109.hg.1 |
TC07003361.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024109.hg.1 |
ENST00000003100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024109.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024110.hg.1 |
TC07003361.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024110.hg.1 |
ENST00000003100.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024110.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008470.hg.1 |
ENST00000419457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008470.hg.1 |
ENST00000419457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201126.hg.1 |
TC0700013584.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000422867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000450723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
ENST00000482924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013584.hg.1 |
NM_001146152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201129.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201129.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201129.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201129.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201129.hg.1 |
TC0700013584.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700201128.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201128.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201128.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201128.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201128.hg.1 |
TC0700013584.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011670.hg.1 |
ENST00000492267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201127.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201127.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201127.hg.1 |
ENST00000003100.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201127.hg.1 |
NM_000786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201127.hg.1 |
TC0700013584.hg.1 |
Clariom D TC |