Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009856.hg.1 |
TC09000767.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009856.hg.1 |
ENST00000303626.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009856.hg.1 |
ENST00000351304.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009856.hg.1 |
ENST00000372080.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009856.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185125.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185125.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185125.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185125.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185125.hg.1 |
TC0900012185.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
ENST00000372080.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012185.hg.1 |
NM_001807.4 |
Refseq |