Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012005.hg.1 |
ENST00000266086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012005.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012005.hg.1 |
TC22000678.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008507.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161261.hg.1 |
TC2200008507.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161261.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161261.hg.1 |
NM_014227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161261.hg.1 |
ENST00000266086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161261.hg.1 |
NM_014227.2 |
Refseq |