Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011940.hg.1 |
ENST00000301202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011940.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011940.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011940.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011940.hg.1 |
TC19000879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011939.hg.1 |
ENST00000301202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011939.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011939.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011939.hg.1 |
TC19000879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011941.hg.1 |
ENST00000301202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011941.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011941.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011941.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011941.hg.1 |
TC19000879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011942.hg.1 |
ENST00000301202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011942.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011942.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011942.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011942.hg.1 |
TC19000879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000610651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
ENST00000610651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169072.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169072.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169072.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169072.hg.1 |
TC1900008862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900169072.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169072.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169072.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000610651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
ENST00000610651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008862.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000610651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
TC1900008862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
ENST00000610651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169076.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
TC1900008862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169074.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
NM_021270.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
TC1900008862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
ENST00000301202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
ENST00000351841.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
ENST00000412608.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
NM_002288.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169075.hg.1 |
NM_021270.4 |
Refseq |