Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017094.hg.1 |
ENST00000003607.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017094.hg.1 |
ENST00000540200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017094.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017094.hg.1 |
TC17001295.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010191.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
ENST00000540200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
ENST00000618887.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
NM_001290145.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
NM_015584.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700179677.hg.1 |
TC1700010191.hg.1 |
Clariom D TC |