Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02044668.hg.1 |
TC02002777.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02044668.hg.1 |
ENST00000295704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02044668.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000474339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015741.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000295704.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000423170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000463188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000473034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
ENST00000497832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
NM_022453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200215561.hg.1 |
TC0200015741.hg.1 |
Clariom D TC |