Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004428.hg.1 |
ENST00000330949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004428.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004428.hg.1 |
TC13000406.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008059.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008059.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004427.hg.1 |
ENST00000330949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004427.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004427.hg.1 |
TC13000406.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008059.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008059.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008059.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154240.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154240.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154240.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154240.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154240.hg.1 |
TC1300008059.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154237.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154237.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154237.hg.1 |
TC1300008059.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154239.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154239.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154239.hg.1 |
ENST00000330949.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154239.hg.1 |
NM_005986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154239.hg.1 |
TC1300008059.hg.1 |
Clariom D TC |