Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003492.hg.1 |
ENST00000376795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003492.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003492.hg.1 |
TC13000336.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003493.hg.1 |
ENST00000376795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003493.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003493.hg.1 |
TC13000336.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007755.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152265.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152265.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152265.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152265.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152265.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152265.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152265.hg.1 |
TC1300007755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152266.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152266.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152266.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152266.hg.1 |
ENST00000376795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152266.hg.1 |
ENST00000602402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152266.hg.1 |
NM_006260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152266.hg.1 |
TC1300007755.hg.1 |
Clariom D TC |