Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001669.3 | NP_001660.2 | 4 - 5 | 541 | 70 | |
NM_009589.3 | NP_033667.2 | 4 - 5 | 541 | 70 | ||
XM_005274514.2 | XP_005274571.1 | 4 - 5 | 511 | 70 | ||
XM_005274515.2 | XP_005274572.1 | 4 - 5 | 511 | 70 | ||
GenBank mRNA | AF160499.1 | - | 4 - 5 | 441 | 70 | |
AK314479.1 | - | 4 - 5 | 505 | 70 | ||
BC020229.1 | - | 469 | 70 | |||
X83572.1 | - | 4 - 5 | 517 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |