Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005143.hg.1 |
TC09000480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005143.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005143.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000375251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
ENST00000466976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008109.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154398.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154398.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154398.hg.1 |
ENST00000375249.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154398.hg.1 |
NM_014282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154398.hg.1 |
TC0900008109.hg.1 |
Clariom D TC |