Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018854.hg.1 |
ENST00000314994.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018854.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018854.hg.1 |
TC16001360.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018855.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018855.hg.1 |
TC16001360.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008907.hg.1 |
ENST00000565150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008907.hg.1 |
ENST00000565150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000567035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
ENST00000567035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188138.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188138.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188138.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188138.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188138.hg.1 |
TC1600011300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600188141.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188141.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188141.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188141.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188141.hg.1 |
TC1600011300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600188140.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188140.hg.1 |
ENST00000567035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188140.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188140.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188140.hg.1 |
ENST00000567035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188140.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188140.hg.1 |
TC1600011300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008907.hg.1 |
ENST00000565150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008907.hg.1 |
ENST00000565150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188142.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188142.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188142.hg.1 |
TC1600011300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000314994.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000567035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
ENST00000567035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011300.hg.1 |
NM_145039.3 |
Refseq |