Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010647.hg.1 |
ENST00000517909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010647.hg.1 |
ENST00000517909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010647.hg.1 |
ENST00000520834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010647.hg.1 |
ENST00000520834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010647.hg.1 |
NR_015374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010647.hg.1 |
NR_015374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005983.hg.1 |
TC08000432.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005983.hg.1 |
ENST00000321870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005983.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007828.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007828.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007828.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007828.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007828.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007828.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007828.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800153850.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800153850.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800153850.hg.1 |
ENST00000321870.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800153850.hg.1 |
NM_152414.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800153850.hg.1 |
TC0800007828.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010647.hg.1 |
ENST00000517909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010647.hg.1 |
ENST00000517909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010647.hg.1 |
ENST00000520834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010647.hg.1 |
ENST00000520834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010647.hg.1 |
NR_015374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010647.hg.1 |
NR_015374.1 |
Refseq |