Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004199.hg.1 |
TC08000297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004199.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004199.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004200.hg.1 |
TC08000297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004200.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004200.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150696.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150696.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150696.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150696.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150696.hg.1 |
TC0800007366.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800150697.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150697.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150697.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150697.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150697.hg.1 |
TC0800007366.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
ENST00000302495.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007366.hg.1 |
NM_153692.3 |
Refseq |