Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006283.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006283.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006283.hg.1 |
TC22000326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006285.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006285.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006285.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006285.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006285.hg.1 |
TC22000326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000465662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000465662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000465662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006284.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006284.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006284.hg.1 |
TC22000326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006286.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006286.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006286.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006286.hg.1 |
TC22000326.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200152570.hg.1 |
TC2200007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152570.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152570.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152570.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152570.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152571.hg.1 |
TC2200007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152571.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152571.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152571.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152571.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000465662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000465662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000465662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007439.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152573.hg.1 |
TC2200007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152573.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152573.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152573.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152573.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152573.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152573.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
TC2200007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
ENST00000498400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152572.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152574.hg.1 |
TC2200007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152574.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152574.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152574.hg.1 |
NM_005297.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152574.hg.1 |
ENST00000249016.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152574.hg.1 |
ENST00000381433.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152574.hg.1 |
NM_005297.3 |
Refseq |