Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012555.hg.1 |
TC0X000942.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012555.hg.1 |
ENST00000359202.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012555.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163939.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163939.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163939.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00163939.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00163939.hg.1 |
TC0X00009340.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
ENST00000359202.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009340.hg.1 |
NM_031894.2 |
Refseq |