Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022968.hg.1 |
TC01001482.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022968.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022968.hg.1 |
ENST00000367763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022968.hg.1 |
ENST00000498166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022968.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022968.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022968.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022969.hg.1 |
TC01001482.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022969.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022969.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177755.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177755.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177755.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177755.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177755.hg.1 |
TC0100010565.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177753.hg.1 |
TC0100010565.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000367763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000465717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000475113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
ENST00000498600.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_001146039.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NM_152281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010565.hg.1 |
NR_027397.1 |
Refseq |