Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005102.hg.1 |
ENST00000291495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005102.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005102.hg.1 |
TC19000377.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007442.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
NM_153221.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
TC1900007442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
ENST00000291495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
ENST00000586018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
ENST00000588333.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900154943.hg.1 |
NM_153221.2 |
Refseq |