Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017085.hg.1 |
TC08001325.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017085.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017085.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017085.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010801.hg.1 |
ENST00000442274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010801.hg.1 |
ENST00000442274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173333.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173333.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173333.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173333.hg.1 |
ENST00000297354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173333.hg.1 |
ENST00000519697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173333.hg.1 |
NM_153225.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173333.hg.1 |
TC0800010800.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010801.hg.1 |
ENST00000442274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010801.hg.1 |
ENST00000442274.1 |
Ensembl |