Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000312553.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000371500.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000395871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000471285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000490650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000490650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000490650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000490650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000490650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
ENST00000618387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199080.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199081.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_001199082.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008341.hg.1 |
NM_153703.4 |
Refseq |